Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Acidaminococcus Fermentans Recombinant D2HGDH (from E. coli)
Supplier: BioVision
AB286018-50UG AB286018-250UG
75838-110EA 1922.88 USD InStock
75838-110 75838-108
Acidaminococcus Fermentans Recombinant D2HGDH (from E. coli)
Enzymes
D-2-hydroxyglutarate (D2HG) level is significantly increased in metabolic diseases and various cancers such as acute myeloid leukemia. Studies suggest that the detection of D2HG serves as a biomarker assay related to IDH (isocitrate dehydrogenase) mutations.
D2HGDH is a special NAD-dependent enzyme which reacts with D2HG specifically and converts D2HG to α-ketoglutarate. D2HGDH is a key enzyme to distinguish between two metabolites, D2HG and L-2-hydroxyglutarate (L2HG), during biomarker assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty