Would you like to log in again?
DiaEasy™ Dialyzer (20 ml) MWCO 6-8 kDa, Biovision
Supplier: BioVision
AB286924-100VIAL AB286924-25VIAL AB286924-10VIAL
10013-378PK 858 USD InStock
10013-378 10013-376 10013-374
DiaEasy™ Dialyzer (20 ml) MWCO 6-8 kDa, Biovision
Protein Purification Protein Purification Dialysis
HTS compatible dialysis tubes with accessories for buffer exchange and sample concentration.
  • Most convenient, user friendly dialysis system on the market. Can be used for:
  • Large-scale protein dialysis such as antibodies and recombinant protein purification.
  • Peptide dialysis as small as 10 amino acids.
  • Virus-particle purification.
  • Removal of contaminating micro molecules.
  • Tissue culture extracts purification.
  • Removal of salts, surfactants, solvents and detergents.
  • Complex formation studies (protein-protein, protein-DNA and protein-RNA).
Biovision’s DiaEasy™ dialyzer (6-8 kDa MWCO) can be used primarily for dialysis or buffer exchanges of sample volumes upto 20 ml. They can also be used for several other purposes as shown below. The DiaEasy™ tube’s membrane is ultra-clean, sulfur and heavy metal free and EDTA treated which makes it suitable for molecular biology experiments. These DiaEasy™ tubes allow rapid, secure and simple loading and recovery. They are very high performance dialysis tubes and are the most convenient, user friendly dialysis system on the market. The sample quality after dialysis can be checked by several assays commonly used for proteins and nucleic acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty