Would you like to log in again?
DiaEasy™ Dialyzer (800 µl) MWCO 6-8 kDa, Biovision
Supplier: BioVision
AB286928-1
10013-476PK 111.32 USD InStock
10013-476
DiaEasy™ Dialyzer (800 µl) MWCO 6-8 kDa, Biovision
Protein Purification Protein Purification Dialysis
HTS compatible dialysis tubes with accessories for buffer exchange and sample concentration.
  • Most convenient, user friendly dialysis system on the market. Can be used for:
  • Dialysis or buffer exchange of small volumes (50-800 µl).
  • Extraction of proteins, RNA, DNA or oligonucleotides (>20 nt) from polyacrylamide, agarose or any gel matrix in any running buffer.
  • Extraction of protein-protein, DNA-protein or RNA-protein complexes.
  • Preparation of protein samples for MALDI-MS.
  • Sample concentration
  • High through put dialysis (using a single beaker with several DiaEasy™ tubes on one floating pad).
Biovision’s DiaEasy™ dialyzer (6-8 kDa MWCO) can combines two modes of action, 1) dialysis or buffer exchange at sample volumes between 50 and 800 µl and 2) electro-elution of macromolecules from polyacrylamide or agarose gel. They can also be used for several other purposes as shown below. This device allows rapid and high performance at either mode and extracts the macromolecules without any contamination. They are ideal for purification of very small quantities of proteins (up to 0.5 µg), for automated protein sequencing, peptide mapping and amino acid analysis. The DiaEasy™ tube’s membrane is ultra-clean, sulfur and heavy metal free and EDTA-treated which makes it suitable for molecular biology experiments. These tubes are autoclaved and bacteria free. The DiaEasy™ tubes allow rapid, secure and simple loading and recovery. They are very high performance dialysis tubes and are the most convenient, user friendly dialysis system on the market. The sample quality after dialysis can be checked by several assays commonly used for proteins and nucleic acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty