Would you like to log in again?
Benchtop Orbital Shaker, Eberbach
Supplier: Eberbach
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
E6130.00
75836-366EA 8300 USD InStock
75836-366
Benchtop Orbital Shaker, Eberbach
Shakers and Mixers Orbital Shakers
This bench top variable speed orbital shaker has been designed to occupy minimal bench space and provide the versatility offered by larger units with its increased load capacity.
  • Digital tachometer and digital countown timer available
  • Motor with 35 mhp
  • Permanently lubricated bearings
  • Rated for continuous operation
The desired rate of rotation is set using the speed control knob on front panel and speed is monitored via tachometer. The digital countdown timer can be selected for timed operation of the unit, unit can also be used for constant operation.
This can be used with optional utility box, flask holders.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty