Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Gauge
Supplier: FISCHER TECHNICAL CO SE
HVG-004
470227-544EA 61.8 USD InStock
470227-544
Vacuum Gauge
Pumps Vacuum Gauges
This bourdon style vacuum gauge features a 2" Dial with a range of 30-0" Hg.
  • Accuracy +-3%
  • Operating Temperature range -40 to 150°F
The connection size is 1/4" NPT and is located on the bottom of the gauge for easy mounting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty