Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tartrazine 1.5% in acetic acid (1,5%)

Supplier: Electron Microscopy Sciences
Synonyms: FD and C Yellow No. 5Acid Yellow 23Filter Yellow

21740
101410-822EA 54.51 USD InStock
101410-822
Tartrazine 1.5% in acetic acid (1,5%)
Tartrazine


Formula: C₁₆H₁₂N₄O₉S₂
MW: 534.36 g/mol
MDL Number: MFCD00148908
CAS Number: 1934-21-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/13/28984913.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty