Would you like to log in again?
Shell Vials, Sun Sri
Supplier: SUN SRi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
500 604 500 634 500 602
66008-234PK 68.73 USD InStock
66008-234 14232-372 66008-270
Shell Vials, Sun Sri
Vials
Vials are made of borosilicate glass, and can be used in Waters WISP™ 96-position autosamplers.
PE snap plug caps feature a tear-away top for easier needle penetration and are kitted unassembled with the appropriate closures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty