Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Snap Top Cap with PTFE/Silicone, PerkinElmer
Supplier: Perkin Elmer
N9303416 N9303419
10136-820PK 73.65 USD InStock
10136-820 10139-724
Snap Top Cap with PTFE/Silicone, PerkinElmer
Closures Chromatography Vial Closures
Snap top closures are designed in a wide variety of sizes and materials based on the sample type and analysis temperature
  • Contaminant free product
  • Excellent resealability
  • Multiple injection capability
Closures are made with specific tolerances that are guaranteed to fit and perform with all type of instruments.
Using a process that bonds silicone/PTFE and otherelastomeric compounds directly into thermoplastic closures eliminates liner fallout.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty