Would you like to log in again?
SP Wilmad-LabGlass Stainless Steel Locking Pinch Clamps, SP Industries
Supplier: SP Industries
LG-1045-118 LG-1045-108 LG-1045-112 LG-1045-114 LG-1045-120 LG-1045-106 LG-1045-104
14007-560EA 128.11 USD InStock
14007-560 14007-550 14007-548 14007-546 14007-556 14007-554 14007-562
SP Wilmad-LabGlass Stainless Steel Locking Pinch Clamps, SP Industries
Clamps General Purpose Clamps
These pinch clamps are for use with ball and socket joints and joints with O-rings.
Models with locking device in addition to spring (Sizes 12A, 18A and 28, 35, 40, 50, 65, and 75) will support considerable weight. The locking device is also necessary when clamps are used with O-rings to furnish needed compression of the O-ring. The size given indicates the nominal diameter of the ball members of the socket joints the clamps will fit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty