Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Ultraviolet UV Light Pen, Sper Scientific

Supplier: Sper Scientific
330005
97035-718EA 62.28 USD InStock
97035-718
Ultraviolet UV Light Pen, Sper Scientific
Pens Ink Pens
UV Light Pen uses new solid-state LED technology to offer a portable source of UV light built into a slim attractive writing ballpoint ink pen.

  • Compact, rugged and portable
  • Wavelength range: 390 to 410 nm
  • Immediate output, no warm up time
  • Light source: High intensity InGaN LED

Luminous Intensity Max. 380 mcd (Milli-candela)
Temperature: –30 to 80 °C
130 x 10 mm (51/8 x 3/8")

Longer battery life eliminates the need for power cords and battery packs. Powered by only four button cell batteries.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty