Would you like to log in again?
Thrombin Substrate III
Supplier: EMD Millipore
605211-25MG 605211-25MG
80054-704EA 108.87 USD InStock
CA80054-704 80054-704
Thrombin Substrate III
Enzymes
Sensitive fluorogenic substrate for the quantitative determination of thrombin.
Following reconstitution, aliquot and freeze (–20 °C). Stock solutions are stable for up to 3 months at –20 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty