Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Choline chloride ≥99% cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

0219463980 0219463991 0219463990
IC0219463980EA 71.31 USD InStock
IC0219463980 IC0219463991 IC0219463990
Choline chloride ≥99% cell culture reagent
Choline chloride
Choline chloride is an organic compound and a quaternary ammonium salt. It is a weak acid. Choline chloride is the salt of the naturally occurring choline, the pre-stage of the neurotransmitter acetylcholine, which is important for mnemonic and thought-processes. Choline occurs naturally in fungi, hop and kingcups and as integral part of lecithin. Choline chloride is a common food additive in animal husbandry
The enzymatic activities of butyrylcholinesterase (BChE) and paraoxonase 1 (PON1), two serum enzymes synthesized by the liver and related with inflammation, were decreased in a sepsis animal model injected with LPS. Choline chloride administered intravenously at 20 mg/kg body weight prevents the LPS-mediated decreases in the activities of these two enzymes.
Choline chloride is used as a source of methyl groups for methyl-transfer reactions.
Physical Appearance: Hard, White Crystalline Powder
pH: Neutral to Litmus
Vapor Pressure: 10 hPa at 20 °C (Lit.)
Partition Coefficient: n octanol/water (logKow): -3.77 at 25 °C (lit.)
Solubility: Soluble in water: (10% aq soln) Clear, Colorless Solution


Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Density: 1.1 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/13/88/36441388.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty