Would you like to log in again?
SuperSep™ Phos-tag™ Precast Gels for XCell SureLock® Mini-Cell, Wako
Supplier: Wako
199-18011 192-18001
103258-494PK 431.54 USD InStock
103258-494 103012-118
SuperSep™ Phos-tag™ Precast Gels for XCell SureLock® Mini-Cell, Wako
Electrophoresis Gels Precast Gels
Ready-to-use electrophorosis precast gels for XCell SureLock® Mini-Cell electrophoresis tank
  • Long-term stability (Stable for 9 months)
  • Basic mechanism as normal SDS-PAGE
  • High reproducibility
  • Safety due to precast gel
It contains zinc as a supplemental metal. It has high storage stability by its neutral gel buffer and ZnCl2 and can result in sharp bands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty