Would you like to log in again?
SP Wilmad-LabGlass Liebig Condensers, SP Industries
Supplier: SP Industries
LG-5150-100 LG-5150-106 LG-5150-104 LG-5150-108
14210-202EA 333.4 USD InStock
14210-202 14209-194 14209-714 14203-054
SP Wilmad-LabGlass Liebig Condensers, SP Industries
Condensers
The top of these Liebig condensers have a Standard Taper outer joint and the bottom has same size (except Model ML-560-702) inner joint, with drip tip.
  • Borosilicate glass
NOTE: Model ML-560-702 has different sized top and bottom joints.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty