Would you like to log in again?
Amino acid mixture 1 mM Minus Methionine
Supplier: Promega Corporation
L9961
PAL9961EA 85.54 USD InStock
PAL9961
Amino acid mixture 1 mM Minus Methionine
Amino acid mixture
Amino Acid Mixture, Complete, is an aqueous solution containing 1mM each of the 20 essential amino acids.
  • 1mM Solution of 20 Essential Amino Acids or Combinations Thereof
  • Amino acid solution for use with Flexi(R), TnT(R) and Rabbit Reticulocyte Lysates, Wheat Germ Extract and E. coli S30 Systems
  • Choose from mix of 20 amino acids or special mixtures indicated
  • Supplied at a concentration 1mM
This mixture is compatible for use in the Flexi Lysate, TNT Lysate and standard Rabbit Reticulocyte Lysate Systems as well as in the Wheat Germ Extract and E. coli S30 Systems.
Amino Acid Mixtures are also available lacking cysteine, methionine and cysteine, leucine or methionine.
Storage: LESS THAN -65°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty