Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hematoxylin 1% in ethanol 95%

Supplier: Electron Microscopy Sciences
Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

26091-01
101411-010EA 63.73 USD InStock
101411-010
Hematoxylin 1% in ethanol 95%
Hematoxylin
Alcoholic stain solutions are available with Congo Red 0.2%, Hematoxylin 1% in 95% alcohol, Iodine 0.5% in 95% alcohol, Iodine 0.5% in 85% alcohol, Iodine 2% in 85% alcohol, Saffron 6%, Thionin 0.5% in 20% alcohol, Aldehyde Fuchsin and Alum Hematoxylin.


Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty