Would you like to log in again?
SP Wilmad-LabGlass Three Hole Mobile Phase Delivery Cap, SP Industries
Supplier: SP Industries
BP-1913-000
14204-158EA 515.16 USD InStock
14204-158
SP Wilmad-LabGlass Three Hole Mobile Phase Delivery Cap, SP Industries
Closures Safety Caps and Safety Waste Caps
The three holes of this mobile phase delivery cap have 1/4"–28 threads in the top with 1/8" holes for 1/8" O.D
  • Body is glass-filled PTFE
  • Screw cap has 45 mm thread
  • Integral pressure relief valve relieves reservoir pressure at 10 psi
Includes three 1/4" -28 ETFE plugs and Viton® O-ring for sealing to the reservoir.
PTFE tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty