Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bacillus licheniformis ALCALASE®

Supplier: EMD Millipore
126741-500ML
80108-030EA 154.62 USD InStock
80108-030
Bacillus licheniformis ALCALASE®
Enzymes
A serine endopeptidase that consists primarily of subtilisin A

  • Density: 1.25 g/mL
  • Activity: ≥0.75 Anson units/ml

One unit is defined as the amount of enzyme that will release 1.0 µmol L-tyrosine from hemoglobin per min at 25°C, pH 7.5.

Suitable for the hydrolysis of proteins.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty