Would you like to log in again?
Specialty Spill Kit: Mercury, Brady Worldwide
Supplier: Brady
148859
76000-814KT 323.6 USD InStock
76000-814
Specialty Spill Kit: Mercury, Brady Worldwide
Spill Control Materials
SPC® spill kit contains surfactant-treated absorbents and contents designed to pick up mercury spills while remaining chemically inert.
  • Acid neutralizer allows for use on even the most aggressive chemicals
Delivery information: Kit includes 10 pads (15" x 19"), 453 g mercury absorbent powder, goggles, scoop and scraper, 1 pair neoprene gloves, 2 disposal bags and instruction sheet
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty