Would you like to log in again?
KIMTECH PURE® A6 Liquid Splash Protection Coverall, Kimberly-Clark
Supplier: Kimberly-Clark
47687 47683
75832-260CS 291.93 USD InStock
CA75832-280 75832-260
KIMTECH PURE® A6 Liquid Splash Protection Coverall, Kimberly-Clark
Disposable Garments
Enhanced liquid barrier protection and comfort for the most critical areas.
  • High-performance laminated film on the front panel, hood, arms, and legs
  • Lightweight, breathable filter material on the back panel to keep workers safe and cool
  • Suitable for use in ISO 7/8 Grade C/D cleanrooms
  • Built-in thumb loops and elastic cuffs
  • Innovative origami-inspired fold design to reduce gowning time
Splash protection is essential when working with blood and other hazardous liquids
The KIMTECH PURE® A6 Liquid Splash Protection Coverall from Kimberly-Clark Professional® offers an ideal combination of protection and comfort through two innovative, low-lint fabrics. KIMTECH PURE® A6 Liquid Splash Protection Coveralls offer seamless protection where it matters most.
Applications include:
ISO 7/8 Grade C/D
Nonsterile Pharmaceutical Manufacturing
Bio-Medical Research
Pharmaceutical Compounding
Biotechnology
But so is delivering worker comfort. The challenge is to provide both in a single suit.
Packaging: 25 individually-packed coveralls per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty