Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
3B Scientific® 4_Stage Prostate Exam Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
1013116
470228-500EA 267.1 USD InStock
470228-500
3B Scientific® 4_Stage Prostate Exam Trainer
Models Human Biology Models
Great size for personal practice!
  • Includes 4 prostate conditions
  • Practice DRE anywhere
  • Compact size and shape for easy storage and use
The Prostate Examination Trainer is used in medical simulation and clinical training for the demonstration and practice of digital rectal examinations (DRE). The trainer includes 4 different prostates to train professional palpation methods (side position or bent forward) and diagnostic skills associated with rectal examinations.
The index finger is inserted into the rectum, facilitating palpation of the rear part of the prostate, where the majority of prostate carcinomas are found. The shape, position and consistency of the prostate are assessed, allowing irregularities, induration and nodes on the otherwise smooth surface of the prostate to be felt.
The following stages are included:
Normal prostate
Advanced prostate carcinoma
Prostate carcinoma at an early stage
Benign prostatic hyperplasia (BPH)
For reference and self-control, the trainer also includes the corresponding transrectal sonographic imaging (transrectal ultrasound, TRUS) for each stage. While in use, the prostate and the sonographic images are hidden and cannot be seen by the trainee.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty