Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia 10% in 85.5% alcohol

Supplier: Electron Microscopy Sciences

10380
101410-616EA 67.7 USD InStock
101410-616
Ammonia 10% in 85.5% alcohol
Ammonia
Alkaline Alcohol formulations are 10% NH4OH in 85.5% alcohol, 1.6% NaCl in 80% alcohol, and 0.2% KOH in 80% alcohol.


Formula: NH₃
MW: 17.03 g/mol
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 1005
ADR: 2,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty