Would you like to log in again?
DNPH Reference Materials, Restek
Supplier: Restek
33082 33091 33074
75835-898EA 56.26 USD InStock
75835-898 75835-148 75835-146
DNPH Reference Materials, Restek
Standards Organic Standards
Caution: *The reported concentrations reflect the amount of aldehyde or ketone in the mixture. The concentration of derivatized aldehyde or ketone is not reported.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty