Would you like to log in again?
FastDNA Spin Kit for Plant and Animal Tissues, MP Biomedicals
Supplier: MP Biomedicals
FastDNA™
116540800
IC116540800EA 658.88 USD InStock
IC116540800
FastDNA Spin Kit for Plant and Animal Tissues, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastDNA™ Spin Kit for Plant and Animals Tissues quickly and efficiently isolates high quality genomic DNA from plant and animal tissues.
  • Optimized green Lysing Matrix D (1.4 mm ceramic beads) 2 mL tubes
  • Silica-based spin filter method for purification
  • For use with FastPrep-24, FastPrep-24 5G and FastPrep-96 Homogenizers
An optimized green Lysing Matrix D, coupled with any FastPrep Instrument, quickly lyses most tissue samples in 40 seconds or less. Following lysis, samples are centrifuged to pellet debris and lysing matrix. DNA is purified from the supernatant with a silica-based procedure using SPIN filters. Eluted DNA is ready for digestion, electrophoresis, PCR and any other desired application.
Ordering information: FastPrep Instrument or alternative high speed disruptor; Microcentrifuge that can freely spin 2.0 mL tubes; Microcentrifuge tubes (2.0 mL and 1.5 mL); Rotator or low-speed vortex are required (sold separately).
Delivery information: All FastDNA™ Spin Kit for Plant and Animals Tissues reagents are shipped ambiently. FastDNA™ Spin Kit for Plant and Animals Tissues reagents are stable at room temperature (15-30 °C).
Packaging: Contains Lysing Matrix D 100x 2.0 mL tubes, 1/4” Ceramic Spheres 100 spheres, Binding Matrix 100 mL, Concentrated SEWS-M 12 mL, DES 25 mL, CLS-VF 90 mL, PPS 25 mL, CLS-TC 110 mL, SPIN Modules 100 each, Catch Tubes 100 each.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty