Would you like to log in again?
Bunsen Burners, Humboldt
Supplier: Humboldt
H-5890
72410-060EA 40.85 USD InStock
72410-060
Bunsen Burners, Humboldt
Heaters Gas Burners
Adjustable burners with threaded needle valves utilize the highquality die-cast bases and provide a precision needle valve to achieve an accurate, adjustable flame.
Overall Height: 156 mm
Mixing Tube OD: 13 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty