Would you like to log in again?
524 Internal Standard/Surrogate Mix (3 components), Restek
Supplier: Restek
30201
RK30201EA 36.86 USD InStock
RK30201
524 Internal Standard/Surrogate Mix (3 components), Restek
Standards Environmental Analysis Standards
Mix contains 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichlorobenzene-d4 (2199-69-1), Fluorobenzene (462-06-6).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty