Would you like to log in again?
L/C Aniline Mix A (3 Components), Restek
Supplier: Restek
31210
RK31210EA 49.47 USD InStock
RK31210
L/C Aniline Mix A (3 Components), Restek
Standards Organic Standards
Contains: 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Caution: Must be calibrated at a level different from the other listed semivolatile compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty