Would you like to log in again?
Organochlorine Pesticide Mix AB #1 (20 Components), Restek
Supplier: Restek
32291
RK32291EA 104.76 USD InStock
RK32291
Organochlorine Pesticide Mix AB #1 (20 Components), Restek
Standards Organic Standards
This mix contains 20 components.
Components are aldrin, α-BHC, β-BHC, δ-BHC, γ-BHC, cis-chlordane, trans-chlordane, 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide (isomer B), methoxychlor.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty