Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Organophosphorus Pesticide Mix, 8270/Appendix IX (9 components), Restek
Supplier: Restek
32419
RK32419EA 124.16 USD InStock
RK32419
Organophosphorus Pesticide Mix, 8270/Appendix IX (9 components), Restek
Standards Organic Standards
This mix contains 9 components.
Components are dimethoate, disulfoton, famphur, methyl parathion, O,O,O-triethyl phosphorothioate, parathion, phorate, sulphotepp, zinophos (thionazine).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty