Would you like to log in again?
USP <467> Calibration Mix #6 (4 Components), Restek
Supplier: Restek
36008
RK36008EA 43.28 USD InStock
RK36008
USP <467> Calibration Mix #6 (4 Components), Restek
Standards Organic Standards
Mix contains: Chloroform (67-66-3), 60 µg/ml; 1,4-Dioxane (123-91-1), 380 µg/ml; Methylene chloride (dichloromethane) (75-09-2), 600 µg/ml; Trichloroethene (79-01-6), 80 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty