Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.5% (by GC-FID) ACS

Supplier: Tedia
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

AR0116-008
89096-138EA 3264.62 USD InStock
89096-138
Acetonitrile ≥99.5% (by GC-FID) ACS
Acetonitrile
  • Meets reagent specifications for testing USP/NF monographs.
  • Packaged under Nitrogen
  • Submicron filtered


Formula: H₃CCN
MW: 41.05 g/mol
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay 99.5% min.
Color (APHA) ≤10
Residue after evaporation ≤0.005%
Titrable acid ≤0.008meq/g
Titrable base ≤0.0006meq/g
Water ≤0.3%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty