Would you like to log in again?
Heat Gun, Light Weight, Cooper Tools
Supplier: Cooper Tools
6966C
37696-762EA 600.17 USD InStock
37696-762
Heat Gun, Light Weight, Cooper Tools
Heaters Hot Air Dryers
This ESD safe heat gun is perfect for repetitive or continuous use in production plants, labs, and electronics service facilities
  • Heavy duty heater and fan components for long service life
  • Light weight for reduced operator fatigue
  • Perfect for shrink tubing and vinyl/T.F.E. epoxy curing, drying, and cooling
  • Three-position switch: HOT – OFF – COOL
Certifications: UL and cUL Listed.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty