Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetone ≥99%, Technical Grade

Supplier: Columbus Chemical Industries, Inc
Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

012000-80601
16650-901CS 273.9 USD InStock
16650-901
Acetone ≥99%, Technical Grade
Acetone


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56 °C (133 °F)
Melting Pt: ‒94 °C (‒137 °F)
Flash Pt: ‒17 °C (1 °F) (closed cup)
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Order Now
Specification Test Results
Odor Mild alcohol. Fruity. Mint-like. Fragrant.
Flammability (solid, gas) Flammable
Upper flammability or explosive limit UEL: 13%
Lower flammability or explosive limit LEL: 2%
Vapor pressure 184 mmHg
Vapor density 2.0
Solubility Completely soluble in water
Partition coefficient: n-octanol/water Log Pow: ‒0.024
Auto-ignition temperature 465 °C (869 °F)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty