Would you like to log in again?
Perfect Fit™ Kevlar Sleeves, Honeywell Safety
Supplier: Honeywell Safety
Perfect Fit™
KVS-2-24TH KVS-2-18TH
46100-537CS 1315.89 USD InStock
46100-537 46100-533
Perfect Fit™ Kevlar Sleeves, Honeywell Safety
Workwear
Perfect Fit™ Kevlar sleeves are made of 100% DuPont™ Kevlar brand fiber provides excellent protection from cut, slash
  • 2-ply construction
  • Thumb slot prevents the sleeve from twisting while in use
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty