Would you like to log in again?
Cole-Parmer® WS-100 Series Stuart Water Stills, Antylia Scientific
Supplier: Antylia Scientific
99293-00
75994-898EA 2202.31 USD InStock
75994-898
Cole-Parmer® WS-100 Series Stuart Water Stills, Antylia Scientific
Water Purification Systems Stills
Save money while maintaining high performance.
  • Two independent thermostats prevent overheating
  • Features a sturdy metal stand for either wall or bench mounting
  • Operate directly from tap water
This basic economical water still is perfect for general laboratory and educational distillations. Borosilicate glass boiler and condenser provide the highest purity distillate. The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate cold distilled water.
Condenser with screw thread connections and plastic 3/8" ID hose barb fittings allow quick and safe tubing change without worrying about breaking the glassware. Easily descale without dismantling the glassware using the built-in acid addition funnel and wastewater drain. Long-life chromium-plated metal heater is protected by twin safety thermostats.
Delivery information: Supplied with support stand with two boiler straps, hose kit, gasket kit, and power cord with stripped ends.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty