Would you like to log in again?
SP Wilmad-LabGlass Iodine Flasks with Stopper, SP Industries
Supplier: SP Industries
LG-7870-100 LG-7870-102 LG-7870-106
14210-378EA 156.29 USD InStock
14210-378 14210-364 14203-142
SP Wilmad-LabGlass Iodine Flasks with Stopper, SP Industries
Flasks Iodine Flasks
These flasks have a funnel top with full length Standard Taper 24/40 joint.
  • Made of borosilicate glass
Complete unit includes flask and closed bottom hollow stopper (LG-10350).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty