Would you like to log in again?
High-Pressure Frit-Type In-Line Filter, Restek
Supplier: Restek
25041
10059-340EA 121.47 USD InStock
10059-340
High-Pressure Frit-Type In-Line Filter, Restek
Filters In-Line Filters
Restek’s high-pressure in-line filter is a stand-alone version of the Trident column protection system, specifically designed for ease of use, low dead-volume, and flexibility.
  • Stand-alone protection system
  • Easy to use
  • Low dead-volume
The filter has a replaceable, PEEK encapsulated 316 stainless steel frit with a surface area of 12 mm². The standard frit with 2.0 µm porosity may be replaced with an optional 0.5 µm porosity frit. Use of this filter can greatly extend column life, thereby reducing costs and saving maintenance time. Tubing ¹/₁₆" OD; connectors–CPI.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty