Would you like to log in again?
SP Wilmad-LabGlass Low Form Gooch Crucibles, SP Industries
Supplier: SP Industries
LG-5480-108
14201-702EA 89.37 USD InStock
14201-702
SP Wilmad-LabGlass Low Form Gooch Crucibles, SP Industries
Crucibles
With sealed-in fritted glass disc, these borosilicate glass low form Gooch crucibles are designed for analytical work involving precipitation, drying, and weighing.
Fine (F), medium (M), and coarse (C) porosities available.
Caution: Avoid sudden temperature changes when heating or cooling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty