Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uranyl acetate 3% in aqueous solution Ready-to-Use

Supplier: Electron Microscopy Sciences
Warning

22400-3
102092-286EA 133.03 USD InStock
102092-286
Uranyl acetate 3% in aqueous solution Ready-to-Use
Uranyl acetate
Pre-made and ready to use uranyl acetate solution are avaialable with concentrations of 1%, 2%, 3%, or 4%.

Other concentrations are available upon request.


Formula: C₄H₈O₆U
Boiling Pt: 100 °C (212 °F)
Storage Temperature: Ambient
CAS Number: 541-09-3
UN: 2910
ADR: 7,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty