Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcian blue 1% in 1% acetic acid

Supplier: Electron Microscopy Sciences
Synonyms: C.I. 74240C.N. AnAn-6Ingrain Blue 1Alcian blue 8 GX

26026-11
101410-876EA 125.03 USD InStock
101410-876
Alcian blue 1% in 1% acetic acid
Alcian blue 8 GX
Alcian Blue Solution, Electron Microscopy Sciences, is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 1.0% aqueous, and 1.0% aqueous in 1% or 3% acetic acid.


Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298.86 g/mol
MDL Number: MFCD00010720
CAS Number: 33864-99-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty