Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutaraldehyde 10% in acetone, anhydrous, EM Grade

Supplier: Electron Microscopy Sciences
Synonyms: 1,5-PentanedialGlutardialdehydeGlutaral

16530
100504-014EA 178.65 USD InStock
100504-014
Glutaraldehyde 10% in acetone, anhydrous, EM Grade
Glutaraldehyde
Anhydrous EM Grade Glutaraldehyde, Specially Prepared 10% Solution, Electron Microscopy Sciences

Rapid-Freezing on a polished metal surface, at the temp. of liquid helium or liquid nitrogen, followed by freeze-substitution in osmium-acetone, has been shown to be a very useful technique for capturing rapid cellular events and for preserving cell ultrastructure.

John M. Robinson and Morris J. Karnovsky, 1991.
"Rapid-freezing Cytochemistry: Preservation of Tubular Lysosomes and Enzyme Activity". J. of Histochem., Cytochem. Vol. 39, No. 6, pp 787-792


Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00007025
CAS Number: 111-30-8
UN: 2927
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty