Would you like to log in again?
DNA Ladder, 100 bp, BioVision
Supplier: BioVision
AB286888-100TEST
76005-240EA 718.09 USD InStock
76005-240
DNA Ladder, 100 bp, BioVision
Electrophoresis Reagents Molecular Weight Markers
100 bp DNA Ladder.
PCR products and double-stranded DNA were digested with appropriate restriction enzymes to completion to yield 17 bands, ranging from 50 bp-1.5 kb, for molecular weight standards in agarose gel electrophoresis. The 200 and 500 bp bands have increased intensity to serve as quick reference points. Approximate mass of each DNA band is provided (for a loading size of 5 μl of the DNA ladder) for approximating the mass of DNA in comparably intense DNA samples of similar size.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty