Would you like to log in again?
Protein A Sepharose, Endotoxin-free, BioVision
Supplier: BioVision
AB289898-5ML AB289898-25ML AB289898-1ML
76005-270EA 3254.78 USD InStock
76005-270 76005-268 76005-266
Protein A Sepharose, Endotoxin-free, BioVision
Chromatography Sorbents HPLC Sorbents
Endotoxin free Protein A Sepharose for High Specific Binding of IgG.
  • High flow rate
  • For high specific binding
  • Endotoxin-free
Endotoxin free Protein A-Sepharose beads are prepared by covalently coupling recombinant Protein A to 6% cross-linked Sepharose beads under proprietary endotoxin-free conditions
Purification of endotoxin-free monoclonal and polyclonal antibodies from culture media, serum, ascites fluid or hybridoma supernatants. Isolation of antibody/antigen complexes in immunoprecipitation experiments, since only the Fc region is involved in antibody binding and the Fab region is available for binding antigen.
BioVision Protein A (Cat. # 6500, Cat. # 6500B) is a genetically engineered protein containing five IgG-binding regions of native Protein A. The cell wall binding region, albumin binding region and other non-specific regions have been eliminated from the recombinant Protein A to ensure the maximum specific IgG binding. The coupling technique is optimized to give a higher binding capacity for IgG & minimum leaching of recombinant Protein A. The IgG binding capacity of Protein A-Sepharose is ≥ 15 mg human or rabbit IgG per ml of wet beads. Protein A-Sepharose beads display high chemical & physical stability as well as high flow rate, hydrophilicity & high gel strength. It can be used for IgG purification and immunoprecipitation. Our Endotoxin-free Protein A-Sepharose also shows ability of reducing/removing certain amount of endotoxin from serum or ascites samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty