Would you like to log in again?
BD BBL™ Mueller Hinton II Agar Prepared Media, 150 mm Plates, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
221994 221800
90006-573PK 56.86 USD InStock
90006-573 90006-472
BD BBL™ Mueller Hinton II Agar Prepared Media, 150 mm Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens.
  • Used primarily for antimicrobial susceptibility testing
  • Ready for immediate use
This line of prepared media is supplied in 150x15mm plates, which are ready for immediate use. Store at 2 to 8°C (36 to 46°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty