Would you like to log in again?
BD BBL™ Mueller Hinton Agar with 5% Sheep Blood Prepared Media, 150 mm Plates, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
221993 221801
90006-475PK 165.08 USD InStock
90006-475 90006-571
BD BBL™ Mueller Hinton Agar with 5% Sheep Blood Prepared Media, 150 mm Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens.
  • Used primarily for antimicrobial susceptibility testing
  • Ready for immediate use
This line of prepared media is supplied in 150 x 15mm plates, which are ready for immediate use. Store at 2 to 8°C (36 to 46°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty