Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acridine orange (hemizinc salt) 0.2% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Basic Orange 143,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt)

26024-02
101410-872EA 77.01 USD InStock
101410-872
Acridine orange (hemizinc salt) 0.2% in aqueous solution
Acridine orange (hemizinc salt)
Electron Microscopy Sciences Acridine Orange Aqueous solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 0.1% aqueous and 0.2% aqueous.


Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
MDL Number: MFCD00081043
CAS Number: 10127-02-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty