Would you like to log in again?
BD BBL™ Stacker™ I Plate™ Columbia CNA Agar, with 5% Sheep Blood/MacConkey II Agar, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™ Stacker™ I Plate™
221601 221600
90006-278PK 219.5 USD InStock
90006-278 90006-277
BD BBL™ Stacker™ I Plate™ Columbia CNA Agar, with 5% Sheep Blood/MacConkey II Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens.
  • Two-section plates containing one or two media formulations
  • Interlocking feature minimizes hazard of sliding plates
  • Ready for immediate use
  • Stackable
These 100x15mm plates feature two sections containing one or two media formulations. Plates will interlock, minimizing the hazard of sliding plates. Plates are ready for immediate use. Store at 2 to 8°C (36 to 46°F). Stackable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty