Would you like to log in again?
BCR® Cleanroom Flat Mop Cart, Berkshire
Supplier: Berkshire
BCR®
BCRMOPCART1
76028-248CS 886.51 USD InStock
76028-248
BCR® Cleanroom Flat Mop Cart, Berkshire
Mops
The Cleanroom Flat Mop Cart is made of high quality stainless steel and is fully autoclavable
  • High quality stainless steel
  • Fully autoclavable
  • Locking casters provide stability during use
The cart includes two 20 liter polypropylene buckets, color-coded blue and red to differentiate between clean and dirty solutions.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty