Would you like to log in again?
Elevator, Sklar
Supplier: Sklar
98-7316 98-7315
75995-550EA 338.85 USD InStock
75995-550 75995-548
Elevator, Sklar
Retractors
Multipurpose instrument for use in micro neurosurgical procedures.
  • Reusable
  • Nonsterile
The elevator is designed to elevate tissue layers to gain clear access to explore the surgical field. The elevator features a straight shaft and comes with either a curved or angled tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty