Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphotungstic acid hydrate 1% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Wolframophosphoric acid hydratePhosphotungstic acid hydrate12-Tungstophosphate hydrateDodecawolframophosphoric acid hydrateTungstophosphoric Acid Hydrate

19502-1
101410-754EA 78.24 USD InStock
101410-754
Phosphotungstic acid hydrate 1% in aqueous solution
Phosphotungstic acid hydrate
Phosphotungstic Acid Aqueous solution is available in a saturated aqueous solution, 0.1% in acetone and 4.67% in 95% alcohol.


Formula: H₃[P(W₃O₁₀)₄]·nH₂O
MW: 2898.07 g/mol
MDL Number: MFCD00011342
CAS Number: 12501-23-4
UN: 3260
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty